Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA018886

Sigma-Aldrich

Anti-BEX3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Bex, Anti-DXS6984E, Anti-HGR74, Anti-NADE, Anti-NGFRAP1, Anti-Brain-expressed X-linked protein 3, Anti-Nerve growth factor receptor-associated protein 1, Anti-Ovarian granulosa cell 130 kDa protein HGR74, Anti-Protein BEX3, Anti-p75NTR-associated cell death executor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

NEEMEQPMQNGEEDRPLGGGEGHQPAGNRRGQARRLAPNFRWAIPNRQINDGMGGDGDDMEIFMEEMREIRRKLRELQLRNCLRILMGELSNHHDHHDEF

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NGFRAP1(27018)

General description

The gene NGFRAP1 (nerve growth factor receptor-associated protein 1) is mapped to human chromosome Xq22.2. It belongs to BEX (brain expressed X-linked) gene family. The protein localizes in the cytoplasm and the nucleus. NGFRAP1 is popularly called as NADE (p75NTR-associated cell death executor).

Immunogen

brain expressed X-linked 3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Nerve growth factor receptor-associated protein 1 (NGFRAP1) interacts with the death domain p75NTR (p75 neurotrophin receptor) and induces apoptosis in response to nerve growth factor.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74413

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Teva Brender et al.
Case reports in genetics, 2015, 453105-453105 (2015-03-20)
Pelizaeus-Merzbacher disease (PMD) is neurodegenerative leukodystrophy caused by dysfunction of the proteolipid protein 1 (PLP1) gene on Xq22, which codes for an essential myelin protein. As an X-linked condition, PMD primarily affects males; however there have been a small number
Jun Mukai et al.
The Journal of biological chemistry, 277(16), 13973-13982 (2002-02-07)
Nerve growth factor (NGF) can induce apoptosis in neural cells via activation of the low affinity neurotrophin receptor p75NTR. NADE (p75NTR-associated cell death executor) is a p75NTR-associated protein that mediates apoptosis in response to NGF by interacting with the death
Enrique Alvarez et al.
Gene, 357(1), 18-28 (2005-06-17)
To better understand the development of ventral mesencephalic dopamine neurons, we performed subtractive hybridization screens to find ventral mesencephalic genes expressed at rat embryonic day 10 when these neurons begin to differentiate. The most commonly identified genes in these screens
Yao Yu et al.
Brain research, 1100(1), 13-20 (2006-06-17)
Expression of human dopamine responsive gene-1 (DRG-1) is up-regulated in response to treatment of dopamine in the rat astrocytes. However, its functions are not clear up to now. In the presented studies, DRG-1 was identified to be a conserved gene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service