Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HPA015484

Sigma-Aldrich

Anti-B4GAT1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-B3GN-T1, Anti-B3GNT1, Anti-B3GNT6, Anti-BETA3GNTI, Anti-iGAT, Anti-iGNT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

EEQDQYFEFFPPSPRSVDQVKAQLRTALASGGVLDASGDYRVYRGLLKTTMDPNDVILATHASVDNLLHLSGLLERWEGPLSVSVFAATKEEAQLATVLAYALSSHCPDMRARVAMHLVCPSRYEAAVPDPREPGEFALLRSCQ

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... B3GNT1(11041)

General description

B3GNT1 (N-acetyllactosaminide β-1,3-N-acetylglucosaminyltransferase) is a glycosyltransferase, which is a member of the B3GNT family. It is a type II transmembrane protein, with its transmembrane region composed of 28 amino acids. In fetal tissues, it has a predominant expression in brain and kidney. In adults, it has a ubiquitous expression, with high expression in brain. This protein is thought to be composed of 415 amino acids. It has a cytoplasmic N-terminal, a transmembrane region, a stalk region, and a Golgi-luminal large catalytic domain.

Immunogen

betaGal beta-1,3-N-acetylglucosaminyltransferase 1 to

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

B3GNT1 (N-acetyllactosaminide β-1,3-N-acetylglucosaminyltransferase) along with UDP-Gal:βGlcNAc β-1,4-galactosyltransferase, polypeptide 1 (B4GALT1), catalyzes the polymerization of N-acetylglucosamine and galactose residues in an alternating manner. It is essential for the formation of i antigen, and along with I-forming β-1,6-N-acetylglucosaminyltransferase (IGnT), it synthesizes I antigen. Mutations in this gene leading to non-glycosylation of α-dystroglycan, result in a severe form of Walker-Warburg syndrome. Mutations in this gene are also linked to congenital muscular dystrophy.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71599

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ranad Shaheen et al.
Neurogenetics, 14(3-4), 243-245 (2013-07-24)
Walker-Warburg syndrome (WWS) is a genetically heterogeneous form of congenital muscular dystrophy with significant brain and ocular involvement. In a multiplex consanguineous family with severe WWS phenotype, autozygome-guided sequencing of previously reported WWS genes was negative. Exome sequencing followed by
Jeremy L Praissman et al.
eLife, 3, doi:10-doi:10 (2014-10-04)
Recent studies demonstrated that mutations in B3GNT1, an enzyme proposed to be involved in poly-N-acetyllactosamine synthesis, were causal for congenital muscular dystrophy with hypoglycosylation of α-dystroglycan (secondary dystroglycanopathies). Since defects in the O-mannosylation protein glycosylation pathway are primarily responsible for
Peter L Lee et al.
Glycobiology, 19(6), 655-664 (2009-03-06)
Poly-N-acetyllactosamine (polyLacNAc) is a linear carbohydrate polymer composed of alternating N-acetylglucosamine and galactose residues involved in cellular functions ranging from differentiation to metastasis. PolyLacNAc also serves as a scaffold on which other oligosaccharides such as sialyl Lewis X are displayed.
K Sasaki et al.
Proceedings of the National Academy of Sciences of the United States of America, 94(26), 14294-14299 (1998-02-07)
The structure and biosynthesis of poly-N-acetyllactosamine display a dramatic change during development and oncogenesis. Poly-N-acetyllactosamines are also modified by various carbohydrate residues, forming functional oligosaccharides such as sialyl Lex. Herein we describe the isolation and functional expression of a cDNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service