Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HPA010986

Sigma-Aldrich

Anti-DEUP1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CCDC67, Anti-FLJ25393, Anti-coiled-coil domain containing 67 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

MKQNKVPRKELPHLKEEIPFELSNLNQKLEEFRAKSREWDKQEILYQTHLISLDAQQKLLSEKCNQFQKQAQSYQTQLNGKKQCLEDSSSEIPRLICDPDPNCEINERDEFIIEKLKSAVNEIALSRNKLQD

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CCDC67(159989)

General description

CCDC67 (coiled-coil domain containing 67) belongs to the CCDC family of proteins, which are characterized by their coiled-coil motif.

Immunogen

deuterosome assembly protein 1 recombinant protein epitope signature tag (PrEST)

Application

Anti-CCDC67 antibody is suitable for immunocytochemistry. Anti-CCDC67 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

CCDC67 (coiled-coil domain containing 67) gene has its promoter methylated in many gastric cell lines and gastric tumors. Hypermethylation leads to suppression of this gene, which might aid in tumorigenesis. Hence, CCDC67 might act as a tumor suppressor gene.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72161

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sung-Joon Park et al.
Carcinogenesis, 33(8), 1494-1501 (2012-05-23)
In this study, the promoter of the gene coiled-coil domain-containing 67 (CCDC67) was found to be frequently methylated in gastric cancer cell lines and in primary gastric tumors, as examined by restriction landmark genomic scanning. In addition, CCDC67 expression was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service