Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

HPA003877

Sigma-Aldrich

Anti-ZFX antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Zinc finger X-chromosomal protein antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

VSLAHCTVPDDVLASDITSASMSMPEHVLTGDSIHVSDVGHVGHVGHVEHVVHDSVVEAEIVTDPLTTDVVSEEVL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZFX(7543)

General description

Zinc finger X-chromosomal protein (also known as ZNF926) is a zinc finger protein encoded by the ZFX gene in humans. It is a member of Zfy family, which is highly conserved in vertebrates. The gene is mapped to human X chromosome.

Immunogen

Zinc finger X-chromosomal protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Zinc finger X-chromosomal protein (ZNF926) controls the self-renewal of embryonic and hematopoietic stem cells as a transcriptional regulator. It is associated with several human cancers, including lymphoma, laryngeal squamous cell carcinoma, prostate cancer and liver cancer. It also contributes to the maintenance of different types of stem cells and the progression of various cancers.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74527

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Jiang et al.
Cancer gene therapy, 19(10), 684-689 (2012-08-18)
Zinc finger protein X-linked (ZFX) is a highly conservative mammalian gene with related functions in cell survival and proliferation. However, there are limited reports on regulation of ZFX as a therapeutic target in cancer treatment. In this study, the expression
Kui Li et al.
International journal of clinical and experimental pathology, 6(11), 2460-2467 (2013-11-15)
ZFX (zinc finger transcription factor, X chromosome-linked) contributes to the maintenance of different types of stem cells and the progression of various cancers. We have previously reported that ZFX knockdown inhibits proliferation of glioma in vitro and in vivo. Since
Claire Nash et al.
Molecular and cellular endocrinology, 471, 1-14 (2017-05-10)
The androgen receptor (AR) is a transcription factor, and key regulator of prostate development and cancer, which has discrete functions in stromal versus epithelial cells. AR expressed in mesenchyme is necessary and sufficient for prostate development while loss of stromal
Keng Po Lai et al.
International journal of cancer, 135(8), 1790-1799 (2014-03-04)
Zinc finger protein X-linked (ZFX) is a zinc finger protein of Zfy family, which is highly conserved in vertebrates. This transcriptional regulator is not only highly expressed in embryonic stem cells (ESC) and hematopoietic stem cells, but is also upregulated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service