Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA003388

Sigma-Aldrich

Anti-MBIP antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-MAP3K12-binding inhibitory protein 1 antibody produced in rabbit, Anti-MAPK upstream kinase-binding inhibitory protein antibody produced in rabbit, Anti-MUK-binding inhibitory protein antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

DCNQENSCARTDAIFTPYPGFKSHVKVSRVVNTYGPQTRPEGIPGSGHKPNSMLRDCGNQAVEERLQNIEAHLRLQTGGPVPRDIYQRIKKLEDKILELEGISPEYFQSVSFSGKRRKVQPPQQNY

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MBIP(51562)

Related Categories

Immunogen

MAP3K12-binding inhibitory protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

MBIP (MAP3K12 binding inhibitory protein 1) gene encodes a protein for a binding partner of MAP3K12 and inhibits its activity. It forms a component of ADA-containing HAT complex (ATAC) and functions in the acetylation of histone H3 and H4. It is identified as a candidate gene for susceptibility to breast cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70622

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Young Ho Lee et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(8), 7699-7705 (2014-05-09)
The aim of this study was to identify candidate single-nucleotide polymorphisms (SNPs) that might affect susceptibility to breast cancer and then elucidate their potential mechanisms and generate SNP-to-gene-to-pathway hypotheses. A genome-wide association study (GWAS) dataset of breast cancer that included
Sevil Zencir et al.
The Biochemical journal, 450(2), 311-320 (2012-11-22)
ADA (alteration/deficiency in activation) 3 is a conserved component of several transcriptional adaptor and HAT (histone acetyltransferase) complexes that regulate RNA polymerase II-mediated gene expression. Within the HAT complexes ADA3 is associated with ADA2 and the HAT GCN5 (general control

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service