Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA001788

Sigma-Aldrich

Anti-PAPOLA antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-PAP antibody produced in rabbit, Anti-Poly(A) polymerase α antibody produced in rabbit, Anti-Polynucleotide adenylyltransferase α antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

EGVKLTALNDSSLDLSMDSDNSMSVPSPTSATKTSPLNSSGSSQGRNSPAPAVTAASVTNIQATEVSVPQVNSSESSGGTSSESIPQTATQPAISPPPKPTVSRVVSSTRLVNPPPRSSGNAATSGNAATKIPTPIVGVK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PAPOLA(10914)

Immunogen

Poly(A) polymerase α recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PAPOLA (poly(A) polymerase α, also referred to as PAP (poly(A) polymerase)) is a key molecule involved in the addition of the poly(A) at the 3′ end of pre-mRNA. It consists of a C-terminal serine/threonine-rich domain with multiple cyclin-dependent kinase (cdk) sites. The C-terminal has target sites for protein–protein interaction with the 25kDa subunit of cleavage factor I (CFI) and with splicing factors U1A and U2AF65 (U2 small nuclear RNA auxiliary factor). It interacts with both G1 and G2-type cyclins and is a substrate for phosphorylation by cyclin-cdk pairs. The activity of PAP can be regulated through phosphorylation by various regulatory proteins during the cell cycle. In the M phase of the cell cycle, the activity of PAP is negatively regulated by cyclin B-cdc2 kinase hyperphosphorylation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70508

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G L Bond et al.
Molecular and cellular biology, 20(14), 5310-5320 (2000-06-24)
We have previously shown that poly(A) polymerase (PAP) is negatively regulated by cyclin B-cdc2 kinase hyperphosphorylation in the M phase of the cell cycle. Here we show that cyclin B(1) binds PAP directly, and we demonstrate further that this interaction
A C Thuresson et al.
Proceedings of the National Academy of Sciences of the United States of America, 91(3), 979-983 (1994-02-01)
We have cloned human poly(A) polymerase (PAP) mRNA as cDNA in Escherichia coli. The primary structure of the mRNA was determined and compared to the bovine PAP mRNA sequence. The two sequences were 97% identical at the nucleotide level, which
Hana Kim et al.
The EMBO journal, 22(19), 5208-5219 (2003-10-01)
Poly(A) polymerase (PAP) is a key enzyme responsible for the addition of the poly(A) at the 3' end of pre-mRNA. The C-terminal region of mammalian PAP carries target sites for protein-protein interaction with the 25 kDa subunit of cleavage factor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service