Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

HPA000845

Sigma-Aldrich

Anti-MCM5 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CDC46 homolog antibody produced in rabbit, Anti-DNA replication licensing factor MCM5 antibody produced in rabbit, Anti-P1-CDC46 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

NRYIIMRSGARQHERDSDRRSSIPITVRQLEAIVRIAEALSKMKLQPFATEADVEEALRLFQVSTLDAALSGTLSGVEGFTSQEDQEMLSRIEKQLKRRFAIGSQVSEHSIIKDFTKQKYPEHAIHKVLQLMLRRGEIQHRMQRKVLY

UniProt accession no.

application(s)

research pathology

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MCM5(4174)

General description

DNA replication licensing factor MCM5 (minichromosome maintenance complex component 5) is a protein encoded by the MCM5 gene in humans and is localized to chromosome region 22q13.1-->q13.2.

Immunogen

DNA replication licensing factor MCM5 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The gene MCM5 (minichromosome maintenance complex component 5) is expressed in various skin diseases and may act as a useful marker to detect cell proliferation in skin tissue sections. MCM5 is essential for transcription activation of Stat1 target genes. It plays a crutial role in DNA replication and is necessary for transcription activation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73405

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Paul et al.
Cytogenetics and cell genetics, 73(4), 317-321 (1996-01-01)
We have isolated the complete cDNA sequence encoding the human homolog of the yeast replication protein Cdc46/Mcm5. The cDNA was used as a probe for chromosomal fluorescence in situ hybridization (FISH), which localized the human Cdc46 gene (CDC46) to chromosome
Houjun Liu et al.
International journal of dermatology, 46(11), 1171-1176 (2007-11-09)
The entire minichromosome maintenance (MCM) family (MCM2-7) play roles in the initiation and elongation of DNA replication. Many studies have demonstrated that MCM proteins may be better indicators of a wide variety of proliferative or cancer cells in malignant tissues.
Marylynn Snyder et al.
Proceedings of the National Academy of Sciences of the United States of America, 102(41), 14539-14544 (2005-10-04)
The eukaryotic minichromosome maintenance (MCM) family of proteins (MCM2-MCM7) is evolutionarily conserved from yeast to human. These proteins are essential for DNA replication. The signal transducer and activator of transcription proteins are critical for the signal transduction of a multitude

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service