Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EP5318000001

Eppendorf® SmartBlock cryo thaw

to hold, 24 x 1.5-2 mL flat bottom tubes, ThermoMixer C & ThermoStat C

Synonym(s):

Eppendorf® SmartBlock for cell thawing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122801
NACRES:
NB.15

feature

to hold 24 x 1.5-2 mL flat bottom tubes

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 5318000001

compatibility

ThermoMixer C & ThermoStat C

General description

Using different vessel sizes within your lab workflow? Flexibility has never been this easy. Eppendorf® offers a broad range of SmartBlocks for tubes from 0.2 mL to 50 mL as well as plates (MTP, DWP, and PCR plates 96 / 384) to use with Eppendorf [M="ThermoMixer"] C and Eppendorf® ThermoStat C. All SmartBlocks are equipped with the outstanding Eppendorf® QuickRelease system that makes the block exchange super fast and easy. Just press the lever on the front of the block and the block can be removed – no tools are needed, done in seconds.

Application

CRYOSTORAGE

Features and Benefits

Optimized block design supports optimal temperature transfer within your sample to receive excellent results
Eppendorf SmartBlock-specific calibration by integrated sensor technology maximizes temperature accuracy and homogeneity
Every SmartBlock is individually inspected in production for temperature accuracy and is delivered incl. s serial-number specific certificate for documentation purpose
Insulated thermoblocks for ergonomic operation - you won't burn your fingers (PhysioCare Concept®)
Automatic block recognition for safe handling
Eppendorf QuickRelease technology enables fast and simple block exchange - no need for tools
SmartBlocks can only be used with ThermoMixer C and ThermoStat C

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
PhysioCare Concept is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service