EP0030118677
Eppendorf® Conical Tubes SnapTec 50
PCR clean, sterile, capacity 45 mL, pack of 8 × 30 ea
Synonym(s):
Conical Centrifuge tube, micro test tubes
About This Item
Recommended Products
material
colorless polypropylene
high-density polyethylene cap (push fit)
sterility
sterile
feature
DNase free
packaging
pack of 8 × 30 ea
manufacturer/tradename
Eppendorf® 0030118677
capacity
45 mL
diam.
30 mm
suitability
suitable for PCR
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Features and Benefits
Cap with flattened and grooved sides supports a user-friendly ergonomic handling, enables a stable upright positioning, and minimizes contamination risk
Eppendorf Conical Tubes SnapTec 50 for fast opening and closing in routine multistep protocols
New! Conical Tubes SnapTec 50 are autoclavable at 121°C for 20 Minutes
Reliable and safe labelling due to a large writing area on the tube and due to a flat and light-colored cap
High g-Safe centrifugation stability up to 17,000 x g
Precise dimensions enable maximum compatibility with centrifuge rotors, mixers and shakers
Optimal sample and pellet visibility
Sterile, pyrogen-, DNase-, RNase-, and DNA-free. Each individual lot is checked and certified for quality and purity
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service