Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EMU161651

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Slc22a15

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

Pricing and availability is not currently available.

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CACCCTGAGTGCAGGTGACCTAGGGGGCAGCATTTATGCCAACCTGGCTCTATCTGGCCTCATAGAGATTCCATCCTACCCTCTCTGCATCTACCTGATTAACCAAAGATGGTTTGGTCGGAAGCGGACACTAGCAGCGTTTCTGTGCCTTGGAGGACTGGCCTGTCTTATTGTGATGTTTCTTCCGGAAAAGAAAGACACAGGCGTGTTTGCAGTGGTCAACAGCCATTCCTTGTCCCTGCTGGGGAAGCTGACCATCAGTGCTGCCTTTAACATTGTTTACATCTACACCTCTGAGCTGTACCCCACTGTCATCAGAAACGTTGGCCTGGGAGCGTGCTCCATGTTCTCCCGAGTCGGTGGGATCATTGCCCCCTTCGTCCCCTCCCTGGTTGAAGGAAGTACAGTGGTCTCTGCCCTTCATCGTGTTTGGAGCCACAGGCCTGACCTCAGGCCTCCTGAGCCTGCTATTGCCAGAGACTCTGAACAGCCCTTTGCTGGAGACATTTTCTG

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
EMU021141EMU208431EMU015611
Gene Information

mouse ... SLC22A15(242126), Slc22a15(242126)

Gene Information

mouse ... SLC22A1(20517), Slc22a1(20517)

Gene Information

mouse ... SLC22A17(59049), Slc22a17(59049)

Gene Information

mouse ... SLC22A18(18400), Slc22a18(18400)

Ensembl | mouse accession no.

ENSMUSG00000033147

Ensembl | mouse accession no.

ENSMUSG00000023829

Ensembl | mouse accession no.

ENSMUSG00000022199

Ensembl | mouse accession no.

ENSMUSG00000000154

product line

MISSION®

product line

MISSION®

product line

MISSION®

product line

MISSION®

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

NCBI accession no.

NM_001039371

NCBI accession no.

NM_009202

NCBI accession no.

NM_021551

NCBI accession no.

NP_001036225

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service