Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EMU149621

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Dtwd2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

Pricing and availability is not currently available. Contact Technical Service

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCCCACCTCTGTCTAGCTTCTGTATTTGCTTTTAAACATGTGCTTCCAATACTTTTTTTTTTTTTTTAAAGAATCTATTCAAAATACGATTAGTCTAGAGTAAGGAGCTGTTGTTTGTAAAATGAGGAAAATTGTGTCTGGGAACATGTTCCCGTCAGTGTTTATAGGTATGTAGGTGTAGATCACTGATGGAATATACCCCGTGAAACTAATGTTTCAGCCTAGTGAATGAAATTATAAGTGTGAAGTAAGTGACACAGAGGGCAGTTTATGTCATTAAGTAGTGTAAGGGAATAGGGAGACGTTACTGTTTACAGTTGTACTCGGACTCAGCTAATGTGACTCACGTCTTCTGTTAGCATCTGAATTCCTGTGGAGACATATAATTTTGAAATAACCTGCATTCTCTATGGGGGATGCTGTGCATAACTTTCAGTGTTCACTGAGCAAGCTAACTGGTGGTAAGAGGTTTTTGATGAAGTTTCTACATGGAAATGACTGAAGTAAATT

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
EMU000791EMU005751EMU007371
esiRNA cDNA target sequence

GGGGCAAGAGGTTGCTATCAAGCAGATTAATTTACAGAAACAACCAAAGAAGGAATTGATCATTAATGAAATTCTGGTGATGAAAGAGTTAAAGAATCCCAACATAGTTAACTTCTTGGACAGTTACCTGGTAGGAGATGAGTTGTTTGTGGTAATGGAGTACCTCGCTGGTGGGTCCCTCACTGATGTTGTAACAGAAACCTGCATGGACGAAGCGCAGATTGCCGCCGTGTGCAGAGAGTGTTTACAGGCGTTGGAGTTTTTACATGCTAATCAAGTGATCCACAGAGACATCAAAAGTGACAATGTGCTTTTGGGAATGGAAGGCTCAGTTAAACTTACTGACTTCGGCTTCTGTGCCCAGATCACTCCTGAACAGAGCAAACGCAGTACTATGGTTGGAACACCGTACTGGATGGCACCAGAGGTGGTGACACGGAAAGCCTATGGTCCCAAAGTTGACATATGGTCTCTGGGCATCATGGCTATCGAGATGGTTGAAGGAGAGC

esiRNA cDNA target sequence

GCAGGTCCTGACCCTTGTAGTCCTGGAGTTCCCCCTGCAACAAGTCGATGGCCTGCTTGACCCAAACCGCTACTGTGCCTTGCTGCTTCCTCTGCTCAAAGCCTGGAGCCCTGTTTTAAGGAACTACATAAAGCGTGCAGCTGACCACTTGGAAGCATTGGCAGCCATTGAGGACTTCTTCTTGGAACATGAAACTCTCGTTACTTCCATGGCCAAGGTCCTGATGGCTTTCTACCAGCTGGAGATCTTGGCTGAGGAAACAATCTTGAGCTGGTTCAGCCAAAGAGACACAACTGACGAAGGCCAGCAGTTACGGAAGAATCAACAGCTGCAGAGGTTCATCCAGTGGCTGAGAGAGGCAGAAGAAGAGTCATCCGAAGATGACTGATGGTGACTGCCA

esiRNA cDNA target sequence

TCAGTGTGACTGGCTGGAAGCTCGGCTGGAGCATTGGCCCTGCTCACCTGATAAAGCATTTACAGACCGTTCAACAGAACAGTTTTTACACGTGTGCGACTCCTTTACAGGCAGCCTTGGCCGAGGCGTTTTGGATCGATATCAAGCGCATGGATGACCCTGAGTGTTACTTTAATTCTCTGCCAAAGGAATTAGAAGTAAAGAGAGATCGGATGGTCCGTTTACTTAACAGCGTTGGCCTGAAACCCATTGTTCCTGACGGGGGTTACTTCATCATTGCTGATGTGTCTTCATTAGGTGCTGACCTCTCGGACATGAACAGCGATGAGCCTTATGACTATAAGTTTGTGAAGTGGATGACGAAACATAAGAAACTGACAGCCATTCCTGTTTCTGCCTTCTGCGACTC

esiRNA cDNA target sequence

GTGGCGAAGCTGTTCTCTTCCATCGTATGTCTATTGCTCTAGCCAATAGCCTTGATGTTCCTAAGAATTTAACCATTACTGACATCAGTAATAAGACAATCACTGTCAATTATGTGGAAGACACCCTGTCTAACTACTACGGCTGGATGGCTCTGCTCTTGGATCAAGAGACCTACTCGCTGCAGTTTGAGAGCCCTTGGATGAACAGATCTCTGCAGTACTCAGCAACGTTTGACAGCTTTGCTCCTGGAAATTACCTCCTGATAATGCACAGGGACCTACCACCTTATCCTGACATCCTCCTCAGATGTGGGAGTCAGGTGGGCCATTCACTTCCATTTCATCCTTTGCCTAGTCAAGACAGAGCCTGTGATTGGTTCTTCAATAGGCAATTGAGGCAGCTCACCTACCT

Ensembl | mouse accession no.

ENSMUSG00000022781

Ensembl | mouse accession no.

ENSMUSG00000003235

Ensembl | mouse accession no.

ENSMUSG00000040213

Ensembl | mouse accession no.

ENSMUSG00000043760

Gene Information

mouse ... PAK2(224105), Pak2(224105)

Gene Information

mouse ... EIF2B5(224045), Eif2b5(224045)

Gene Information

mouse ... CCBL2(229905), Ccbl2(229905)

Gene Information

mouse ... PKHD1(241035), Pkhd1(241035)

NCBI accession no.

NM_177326

NCBI accession no.

NM_172265

NCBI accession no.

NP_776124

NCBI accession no.

NM_153179

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service