Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EMU028161

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Cdc2a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ACGAATCTCTGGCAAAATGGCCCTGAAGCACCCGTACTTTGATGACTTGGACAATCAGATTAAGAAGATGTAGCCCTCTGGATGGATGTCCCTGTCTGCTGGTCGTAGGGGAAGATCGTGTTGTTTACCGTTGGCTCTCTTCCTGTCTTGTATAGTTTTCTTTGTTTGTAAACTGTCATCTGGACTTTTCTTAATTTCCTACGTATAACTTAATTAACATGTAAATATTATTCCATATGAATTTAAATATAATTCTGTATATGTGCAGATGTCACTGTGGTGGCTGTTAATTACTATAACACAAGTGTTAATTACTACAACATAAGACTTGAGTCTCCCTAGACTTCCCAGCAGCCATTCCTGCAGCTCGGAGCACAGTTGAAGGAGCTGAGCTCAGGCCTCGTGATGCTTTCAAGTGCCTCCGTGTTCTGGATATATATGATTCCTGGTCAGTTTCTTGCC

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Griselda Vallejo et al.
PloS one, 9(5), e97311-e97311 (2014-05-27)
Although non-genomic steroid receptor pathways have been studied over the past decade, little is known about the direct gene expression changes that take place as a consequence of their activation. Progesterone controls proliferation of rat endometrial stromal cells during the
Gunjan Guha et al.
PloS one, 10(5), e0125322-e0125322 (2015-05-06)
Pactamycin, although putatively touted as a potent antitumor agent, has never been used as an anticancer drug due to its high cytotoxicity. In this study, we characterized the effects of two novel biosynthetically engineered analogs of pactamycin, de-6MSA-7-demethyl-7-deoxypactamycin (TM-025) and
Qinghua Xi et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 36(7), 4939-4948 (2015-04-26)
Overexpression of cyclin-dependent kinase 1 (CDK1) has been noted to correlation with several human cancers. However, the effects of CDK1 on ovarian cancer development remain unclear. The aim of this study was to examine the effect of CDK1 and related
Qing Xia et al.
International journal of oncology, 44(3), 735-744 (2014-01-01)
Breast cancer is one of the most common malignancies in women. Approximately 15% of the patients belong to the triple-negative breast cancer (TNBC) group, and have the disadvantage of not benefiting from currently available receptor-targeted systemic therapies. Some cancers in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service