description
Powered by Eupheria Biotech
Quality Level
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
CCATACCTGCTAGGCACCATGGCTGGGGGCGCAGCGGATTGCAGCTTCTGGGAACGGCTGTTGGCTCGGCAATGTCGAATCTATGAGCTTCGAAATAAGGAACGCATCTCTGTAGCAGCTGCCTCCAAACTGCTTGCCAACATGGTGTATCAGTACAAAGGCATGGGGCTGTCCATGGGCACCATGATCTGTGGCTGGGATAAGAGAGG
Ensembl | human accession no.
NCBI accession no.
shipped in
ambient
storage temp.
−20°C
Gene Information
human ... PSMB5(5693) , PSMB5(5693)
Related Categories
General description
MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Xiaodong Feng et al.
Cancer cell, 35(3), 457-472 (2019-02-19)
Activating mutations in GNAQ/GNA11, encoding Gαq G proteins, are initiating oncogenic events in uveal melanoma (UM). However, there are no effective therapies for UM. Using an integrated bioinformatics pipeline, we found that PTK2, encoding focal adhesion kinase (FAK), represents a
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service