Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU121691

Sigma-Aldrich

MISSION® esiRNA

targeting human ATAD2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGAAACCTACCACCGGACACGTGCTTTAAGATCTTTGAGAAAAGATGCACAGAATTCTTCAGATTCTAGTTTTGAGAAGAATGTGGAAATAACGGAGCAACTTGCTAATGGCAGGCATTTTACAAGGCAGTTGGCCAGACAGCAGGCTGATAAAAAAAAAGAAGAGCACAGAGAAGACAAAGTGATTCCAGTTACTCGGTCATTGAGGGCTAGAAACATCGTTCAAAGTACAGAACACTTACATGAAGATAATGGTGATGTTGAAGTGCGTCGAAGTTGTAGGATTAGAAGTCGTTATAGTGGTGTAAACCAGTCCATGCTGTTTGACAAACTTATAACTAACACTGCTGAAGCTGTACTTCAAAAAATGGATGACATGAAGAAGATGCGTAGACAGCGAATGA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Hong et al.
Neoplasma, 63(6), 846-855 (2016-08-28)
Colorectal cancer is one of the most common malignant tumors with a high rate of distant metastasis, postoperative recurrence and mortality. ATPase family AAA domain-containing protein 2 (ATAD2), a member of ATPase family, is highly expressed in various cancers, including
Le Zheng et al.
Oncology reports, 33(5), 2337-2344 (2015-03-31)
The ATPase family AAA domain-containing protein 2 (ATAD2) is associated with many cellular processes, such as cell proliferation, invasion and migration. However, the molecular biological function of the ATAD2 gene in cervical cancer is unclear. The purpose of this study
Wen-Jing Lu et al.
Oncotarget, 6(39), 41722-41735 (2015-10-27)
The ATPase family, AAA domain containing 2 (ATAD2) is highly expressed in multiple cancers. We aim to understand the clinical and biological significance of ATAD2 over-expression in hepatocellular carcinoma (HCC), as a means to validate it as a therapeutic target

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service