Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU102241

Sigma-Aldrich

MISSION® esiRNA

targeting human SELENOM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GACAGCTGAACCGCCTAAAGGAGGTGAAGGCTTTCGTCACGCAGGACATTCCATTCTATCACAACCTGGTGATGAAACACCTCCCTGGGGCCGACCCTGAGCTCGTGCTGCTGGGCCGCCGCTACGAGGAACTAGAGCGCATCCCACTCAGTGAAATGACCCGCGAAGAGATCAATGCGCTAGTGCAGGAGCTCGGCTTCTACCGCAAGGCGGCGCCCGACGCGCAGGTGCCCCCCGAGTACGTGTGGGCGCCCGCGAAGCCCCCAGAGGAAACTTCGGACCACGCTGACCTGTAGGTCCGGGGGCGCGGCGGAGCTGGGACCTACCTGCCTGAGTCCTGGAGACAGAATGAAGCGCTC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Guerriero et al.
European journal of histochemistry : EJH, 58(4), 2433-2433 (2015-01-13)
Selenium is an essential trace mineral of fundamental importance to human healthy and exerts its biological function through selenoproteins. In particular, Selenoprotein M (SELM) is located in the endoplasmic reticulum and contains the common redox motif of cysteine-X-X-selenocysteine type. It

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service