Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV48756

Sigma-Aldrich

Anti-NEK6 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-NIMA (never in mitosis gene a)-related kinase 6, Anti-SID6-1512

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

36 kDa

species reactivity

rat, human, bovine, guinea pig, dog, horse, rabbit, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NEK6(10783)

General description

NEK6 codes for NIMA-related kinase 6 that is involved in cytokinesis and mitotic spindle formation. It is activated by Nek9 and is known to phosphorylate Eg5 during cell signaling cascades.
Rabbit Anti-NEK6 antibody recognizes canine, chicken, zebrafish, human, mouse, rat, bovine, and pig NEK6.

Immunogen

Synthetic peptide directed towards the N terminal region of human NEK6

Application

Rabbit Anti-NEK6 antibody is suitable for western blot concentration of 1μg/ml.

Biochem/physiol Actions

The Aspergillus nidulans ′never in mitosis A′ (NIMA) gene encodes a serine/threonine kinase that controls initiation of mitosis. NIMA-related kinases (NEKs) are a group of protein kinases that are homologous to NIMA. Evidence suggests that NEKs perform functions similar to those of NIMA.The Aspergillus nidulans ′never in mitosis A′ (NIMA) gene encodes a serine/threonine kinase that controls initiation of mitosis. NIMA-related kinases (NEKs) are a group of protein kinases that are homologous to NIMA. Evidence suggests that NEKs perform functions similar to those of NIMA.[supplied by OMIM]. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

Sequence

Synthetic peptide located within the following region: FRCSLADFQIEKKIGRGQFSEVYKATCLLDRKTVALKKVQIFEMMDAKAR

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph Rapley et al.
Journal of cell science, 121(Pt 23), 3912-3921 (2008-11-13)
Nek6 and Nercc1 (also known as Nek9) belong to the NIMA family of protein kinases. Nercc1 is activated in mitosis, whereupon it binds, phosphorylates and activates Nek6. Interference with Nek6 or Nercc1 in mammalian cells causes prometaphase-metaphase arrest, and depletion
Laura O'Regan et al.
Molecular and cellular biology, 29(14), 3975-3990 (2009-05-06)
Nek6 and Nek7 are members of the NIMA-related serine/threonine kinase family. Previous work showed that they contribute to mitotic progression downstream of another NIMA-related kinase, Nek9, although the roles of these different kinases remain to be defined. Here, we carried
Christopher Belham et al.
The Journal of biological chemistry, 278(37), 34897-34909 (2003-07-04)
The Nek family of protein kinases in humans is composed of 11 members that share an amino-terminal catalytic domain related to NIMA, an Aspergillus kinase involved in the control of several aspects of mitosis, and divergent carboxyl-terminal tails of varying

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service