Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

AV32564

Sigma-Aldrich

Anti-FOXP1 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Forkhead box P1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

75 kDa

species reactivity

horse, mouse, human, rabbit, rat, guinea pig

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FOXP1(27086)

General description

FOXP1 is a transcription regulator that is expressed in B cells. High levels of FOXP1 expression have been associated with poor prognosis in patients with diffuse large B-cell lymphomas (DLBCLs). Chromosomal aberrations in FOXP1 gene have been linked to mucosa-associated lymphoid tissue (MALT) lymphoma.
Rabbit Anti-FOXP1 antibody recognizes bovine, canine, chicken, human, mouse, and rat FOXP1.

Immunogen

Synthetic peptide directed towards the N terminal region of human FOXP1

Application

Rabbit Anti-FOXP1 antibody can be used for IHC (4-8μg/ml) and western blotting (0.5μg/ml) applications.

Biochem/physiol Actions

FOXP1 belongs to subfamily P of the forkhead box (FOX) transcription factor family. Forkhead box transcription factors play important roles in the regulation of tissue- and cell type-specific gene transcription during both development and adulthood. Forkhead box P1 protein contains both DNA-binding- and protein-protein binding-domains. This gene may act as a tumor suppressor as it is lost in several tumor types and maps to a chromosomal region (3p14.1) reported to contain a tumor suppressor gene(s).

Sequence

Synthetic peptide located within the following region: MIPTELQQLWKEVTSAHTAEETTGNNHSSLDLTTTCVSSSAPSKTSLI

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sharon L Barrans et al.
Blood, 104(9), 2933-2935 (2004-07-09)
FOXP1 (Forkhead box-P1) is a winged-helix transcription factor that is differentially expressed in resting and activated B cells. FOXP1 expression has been demonstrated in a subset of diffuse large B-cell lymphomas (DLBCLs) and is more common in the nongerminal center
B Streubel et al.
Leukemia, 19(4), 652-658 (2005-02-11)
The three chromosomal translocations t(11;18)(q21;q21), t(14;18)(q32;q21), and t(1;14)(p22;q32) are associated with MALT lymphoma. In a case of MALT lymphoma of the thyroid, we observed t(3;14)(p14.1;q32) by cytogenetic analysis. Fluorescence in situ hybridization studies showed that the immunoglobulin heavy chain locus
Guoxiao Guo et al.
Oncology letters, 12(2), 1554-1558 (2016-07-23)
The aim of the present study was to investigate the expression and function of forkhead box protein 3 (FOXP3) in gastric cancer using a rat model. A total of 92 Wistar rats were divided into two groups: An experimental group
Miaoxia He et al.
Gastric cancer : official journal of the International Gastric Cancer Association and the Japanese Gastric Cancer Association, 17(3), 431-441 (2013-11-16)
Mucosa-associated lymphoid tissue (MALT) lymphoma and diffuse large B-cell lymphoma (DLBCL), which are the two most common types of gastric lymphomas, have different clinicopathological features and molecular characteristics with distinct clinical outcomes. Tumor suppressor miR-34a connects the p53 network with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service