Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV30206

Sigma-Aldrich

Anti-RAD17 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-CCYC, Anti-HRAD17, Anti-R24L, Anti-RAD17 homolog (S. pombe), Anti-RAD17Sp, Anti-Rad24

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

66 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAD17(5884)

General description

RAD17 is a checkpoint protein that has been implicated in replication stress, checkpoint termination and genotoxic stress.
Rabbit Anti-RAD17 antibody recognizes bovine, canine, human, mouse, and rat RAD17.

Immunogen

Synthetic peptide directed towards the C terminal region of human RAD17

Application

Rabbit Anti-RAD17 antibody can be used for western blot (1.25μg/ml) assays.

Biochem/physiol Actions

RAD17 is highly similar to the gene product of Schizosaccharomyces pombe rad17, a cell cycle checkpoint gene required for cell cycle arrest and DNA damage repair in response to DNA damage. This protein shares strong similarity with DNA replication factor C (RFC), and can form a complex with RFCs. This protein binds to chromatin prior to DNA damage and is phosphorylated by ATR after the damage. This protein recruits the RAD1-RAD9-HUS1 checkpoint protein complex onto chromatin after DNA damage, which may be required for its phosphorylation.The protein encoded by this gene is highly similar to the gene product of Schizosaccharomyces pombe rad17, a cell cycle checkpoint gene required for cell cycle arrest and DNA damage repair in response to DNA damage. This protein shares strong similarity with DNA replication factor C (RFC), and can form a complex with RFCs. This protein binds to chromatin prior to DNA damage and is phosphorylated by ATR after the damage. This protein recruits the RAD1-RAD9-HUS1 checkpoint protein complex onto chromatin after DNA damage, which may be required for its phosphorylation. The phosphorylation of this protein is required for the DNA-damage-induced cell cycle G2 arrest, and is thought to be a critical early event during checkpoint signaling in DNA-damaged cells. Eight alternatively spliced transcript variants of this gene, which encode four distinct proteins, have been reported.

Sequence

Synthetic peptide located within the following region: PTQATVPETWSLPLSQNSASELPASQPQPFSAQGDMEENIIIEDYESDGT

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xin Wang et al.
Molecular cell, 23(3), 331-341 (2006-08-04)
The ATR-mediated checkpoint is not only critical for responding to genotoxic stress but also essential for cell proliferation. The RFC-related checkpoint protein Rad17, a phosphorylation substrate of ATR, is critical for ATR-mediated checkpoint signaling and cell survival. Here, we show
Liyong Zhang et al.
The EMBO journal, 29(10), 1726-1737 (2010-04-29)
Recent studies have shown a critical function for the ubiquitin-proteasome system (UPS) in regulating the signalling network for DNA damage responses and DNA repair. To search for new UPS targets in the DNA damage signalling pathway, we have carried out

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service