Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

TX0220

Supelco

n-Tetradecane

Synonym(s):

Tetradecane

Sign Into View Organizational & Contract Pricing


About This Item

vapor pressure

1.55 Pa ( 25 °C)

Quality Level

assay

≥99% (GC)

form

liquid

color

APHA: ≤10

mp

6 °C

transition temp

flash point 102 °C

solubility

<0.0001 g/L

storage temp.

room temp

Analysis Note

Assay (GC):99% min
Color (APHA):10 max
Form:Clear liquid
Infrared Spectrum:Conforms to standard

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Asp. Tox. 1

supp_hazards

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

215.6 °F - closed cup

flash_point_c

102 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service