Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

SCC465

Sigma-Aldrich

BT-RMS Brain-tropic RET Melanoma-sorted Mouse Cell Line

Mouse

Synonym(s):

Q61R NRAS melanoma cell line, SK-MEL-147, human melanoma cell line, human melanoma cells, metastatic melanoma cell line

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

Product Name

BT-RMS Brain-tropic RET Melanoma-sorted Mouse Cell Line,

biological source

mouse

Quality Level

packaging

vial of ≥1X10⁶ vial

manufacturer/tradename

Millipore

technique(s)

cell culture | mammalian: suitable

shipped in

liquid nitrogen

storage temp.

−196°C

General description

The mouse brain-tropic RET melanoma-sorted (BT-RMS) cell line is a derivative of the Ret-melanoma sorted cell line (RMS, Cat. No. SCC462). The RMS parental cells were isolated from a spontaneously occurring skin tumor in RET transgenic mice. SCC465 cells were derived from brain tumors formed after intracardiac injection of Ret malanoma-sorted (RMS) cells.

Application

  • Each vial contains >1X106 viable cells.
  • BT-RMS cells are verified to be of mouse origin and negative for human, rat, Chinese hamster, Golden Syrian hamster, and non-human primate interspecies contamination, as assessed by a Contamination Clear panel by Charles River Animal Diagnostic Services
  • Cells tested negative for infectious diseases against a Mouse Essential CLEAR panel by Charles River Animal Diagnostic Services.
  • Cells tested negative for mycoplasma.

Features and Benefits

SCC465, the BT-RMS brain-tropic RET melanoma-sorted mouse cell line, expresses mCherry and exhibits increased incidence and shortened timeline of brain metastasis post-subdermal injection.​

Target description

The RET oncogene was originally created by an accidental in-vivo recombination of two human genes, Ret and RFP (the Ret-finger protein then, known today as TRIM27), during a transformation of the NIH3T3 cells with human lymphoma DNA.1 The founder Ret tran

Storage and Stability

BT-RMS cells should be stored in liquid nitrogen until use. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Other Notes

This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact l

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service