MABT1583
Anti-Filaggrin Antibody, clone AHF2
Synonym(s):
FLG
About This Item
biological source
mouse
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
AHF2, monoclonal
mol wt
calculated mol wt 435.17 kDa
observed mol wt ~N/A kDa
purified by
ion-exchange chromatography
species reactivity
human
packaging
antibody small pack of 100
technique(s)
ELISA: suitable
electron microscopy: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable
isotype
IgG1κ
epitope sequence
Unknown
Protein ID accession no.
UniProt accession no.
storage temp.
-10 to -25°C
Gene Information
human ... FLG(2312)
Specificity
Immunogen
Application
Evaluated by Immunohistochemistry (Paraffin) in Human skin tissue sections.
Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution of this antibody detected Profilaggrin in Human skin tissue sections.
Tested Applications
Western Blotting Analysis: A representative lot detected Profilaggrin in Western Blotting application. (Sebbag, M., et al. (1995). J Clin Invest.95(6):2672-9; Simon, M., et al. (1995). Clin Exp Immunol. 100(1):90-8; Simon, M., et al. (1995). J Invest Dermatol. 105(3):432-7; Simon, M., et al. (1996). Biochem J. 317(Pt1)(Pt1):173-7).
ELISA Analysis: A representative lot detected Profilaggrin in ELISA application. (Miedouge, M., et al. (2001). Tumour Biol. 22(1):19-26).
Immunohistochemistry Applications: A representative lot detected Profilaggrin in Immunohistochemistry application. (Guy, R., et al. (1998). Dermatology. 196(1):32-7; Chavanas, S., et al. (2006). J Dermatol Sci. 44(2):63-72).
Immunofluorescence Analysis: A representative lot detected Profilaggrin in Immunofluorescence application. (Simon, M., et al. (1995). J Invest Dermatol.;105(3):432-7; Chavanas, S., et al. (2006). J Dermatol Sci.;44(2):63-72).
Electron Microscopy: A representative lot detected Profilaggrin in Electron Microscopy applications (Simon, M., et al. (1995). Clin Exp Immunol. 100(1):90-8; Simon, M., et al. (1995). J Invest Dermatol. 105(3):432-7).
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Physical form
Reconstitution
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service