Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABT1229

Sigma-Aldrich

Anti-PDCD6-interacting protein Antibody, clone 12H1.2

clone 12H2.1, from mouse

Synonym(s):

Programmed cell death 6-interacting protein, PDCD6-interacting protein, ALG-2-interacting protein 1, ALG-2-interacting protein X, Hp95

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

12H2.1, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... PDCD6IP(10015)

General description

Programmed cell death 6-interacting protein (UniProt: Q8WUM4; also known as PDCD6-interacting protein, PDCD6IP, ALG-2-interacting protein 1, ALG-2-interacting protein X, Hp95) is encoded by the PDCD6IP (also known as AIP1, ALIX, KIAA1375) in human. PDCD6IP is a multifunctional protein that interacts with PDCD6 in a calcium-dependent manner. It is involved in endocytosis, multivesicular body biogenesis, membrane repair, cytokinesis, apoptosis and maintenance of tight junction integrity. It is a class E VPS protein that is involved in concentration and sorting of cargo proteins of the multivesicular body (MVB) for incorporation into intralumenal vesicles (ILVs) that are generated by invagination and scission from the limiting membrane of the endosome. It also serves as an adapter for a subset of ESCRT-III proteins, such as CHMP4, to function at distinct membranes. It is reported to play a role in the regulation of both apoptosis and cell proliferation and its overexpression blocks apoptosis. Three isoforms of PDCD6IP have been described that are produced by alternative splicing.

Specificity

Clone 12H2.1 detects Programmed cell death 6-interacting protein in human cells. It targets an epitope within 126 amino acids from the C-terminal region.

Immunogen

GST/His-tagged recombinant fragment corresponding to 126 amino acids from human Programmed cell death 6-interacting protein (PDCD6).

Application

Anti-PDCD6-interacting protein, clone 12H1.2, Cat. No. MABT1229 is a mouse monoclonal antibody that detects Programmed cell death 6-interacting protein and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Western Blotting Analysis: 1 µg/mL from a representative lot detected PDCD6-interacting protein in Jurkat cell lysate.

Immunohistochemistry Analysis: A 1:2,000 dilution from a representative lot detected PDCD6-interacting protein in human pancreas, human prostate cancer, and human cervical cancer tissues.

Quality

Evaluated by Western Blotting in TF-1 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected PDCD6-interacting protein in TF-1 cell lysate.

Target description

~96 kDa observed; 96.02 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service