MABS813
Anti-MAP2K2/MEK2 Antibody
mouse monoclonal, 7F5
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Product Name
Anti-MAP2K2, clone 7F5 Antibody,
antibody form
ascites fluid
Quality Level
clone
7F5
monoclonal
purified by
(unpurified)
species reactivity
mouse, mouse, rat, human
concentration
(Please refer to lot specific datasheet.)
technique(s)
flow cytometry: suitable
immunocytochemistry: suitable
western blot: suitable
isotype
IgG1
UniProt accession no.
General description
Purified recombinant fragment of human MAP2K2 expressed in E. Coli.
Application
Immunocytochemistry Analysis (ICC): A 1:200-1,000 dilution from a representative lot detected MAP2K2 in HeLa cells.
Flow Cytometry Analysis (FC): A 1:200-400 dilution from a representative lot detected MAP2K2 in HeLa cells.
Flow Cytometry Analysis (FC): A 1:200-400 dilution from a representative lot detected MAP2K2 in HeLa cells.
Quality
Evaluated by Western Blotting in PC-12, Jurkat, HeLa and NIH/3T3 cell lysates.
Western Blotting Analysis (WB): A 1:500-2,000 dilution of this antibody detected MAP2K2 in PC-12, Jurkat, HeLa and NIH/3T3 cell lysates.
Western Blotting Analysis (WB): A 1:500-2,000 dilution of this antibody detected MAP2K2 in PC-12, Jurkat, HeLa and NIH/3T3 cell lysates.
Physical properties
~48 kDa observed. Uncharacterized bands may be observed in some lysate(s).
Physical form
Mouse monoclonal IgG1ascitic fluid containing 0.03% sodium azide.
Storage and Stability
Stable for 1 year at -20°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service