MABS1805M
Anti-Dach1 Antibody, clone 1F6.1
clone 1F6.1, from mouse
Synonym(s):
Dachshund homolog 1
About This Item
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
1F6.1, monoclonal
species reactivity
mouse, human
packaging
antibody small pack of 25 μg
technique(s)
immunohistochemistry: suitable (paraffin)
western blot: suitable
isotype
IgG2aκ
NCBI accession no.
UniProt accession no.
target post-translational modification
unmodified
Gene Information
human ... DACH1(1602)
Related Categories
General description
Specificity
Immunogen
Application
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected Dach1 in Neuro-2a cell lysate.
Signaling
Quality
Western Blotting Analysis: 0.5 µg/mL of this antibody detected Dach1 in 10 µg of SHSY-5Y cell lysate.
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service