Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MABN2644

Sigma-Aldrich

ANTI-TAU (MAPT) Antibody

mouse monoclonal, 4B5

Synonym(s):

MAPTL, MTBT1, Neurofibrillary tangle protein, Paired helical filament-tau (PHF-tau), TAU

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

Product Name

Anti-Tau (MTBR3R) Antibody, clone 4B5,

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

4B5, monoclonal

mol wt

calculated mol wt 78.93 kDa
observed mol wt ~60 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

Gene Information

human ... MAPT(4137)

Specificity

Clone 4B5 is a mouse monoclonal antibody that detects Tau. It targets an epitope within 27 amino acids from the C-terminal half. It preferentially reacts with 3R tau isoforms, however, it also displays slight reactivity with 4R tau isoforms.

Immunogen

KLH-conjugated linear peptide corresponding to 27 amino acids from the C-terminal half of human Tau.

Application

Quality Control Testing

Evaluated by Western Blotting with recombinant, human Tau, isoform 2N3R.

Western Blotting Analysis: A 1:500 dilution of this antibody detected recombinant, human Tau protein, isoform 2N3R.

Tested Applications

Western Blotting Analysis: A representative lot detected Tau (MTBR3R) in Western Blotting application (Kanaan, N.M., et al. (2020). Nat Commun. 11(1):2809).

ELISA Analysis: A representative lot detected Tau (MTBR3R) in ELISA application (Kanaan, N.M., et al. (2020). Nat Commun. 11(1):2809).

ELISA Analysis: A representative lot detected Tau recombinant protein, isoform 2N4R (blue line) and 2N3R (red line) (Data courtesy of Nicholas Kanaan, Ph.D., Michigan State University, USA).

Immunohistochemistry Applications: A representative lot detected Tau (MTBR3R) in Immunohistochemistry applications in human temporal cortex tissues from an Alzheimer s disease patient (Data courtesy of Nicholas Kanaan, Ph.D., Michigan State University, USA).

Western Blotting Analysis: A 1:500 dilution of this antibody detected recombinant, human Tau protein, isoform 2N4R.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Microtubule-associated protein tau (UniProt: P10636; also known as Neurofibrillary tangle protein Paired helical filament-tau (PHF-tau)) is encoded by the MAPT (also known as MAPTL, MTBT1, TAU) gene (Gene ID: 4137) in human. Tau is a highly abundant protein found predominantly in the nervous system. Its primary role is to stabilize and support microtubule assembly, which are essential for maintaining the structure and function of nerve cells. It is present throughout the brain, particularly in regions involved in memory and cognition. MAPT gene consists of multiple exons that undergo alternative splicing, resulting in the generation of different isoforms that differ in the number and arrangement of repeats in the microtubule-binding domain. The expression of the MAPT transcripts varies and depends on the stage of neuronal maturation and neuron type. Aberrant aggregation and accumulation of tau protein have been linked to several neurodegenerative diseases collectively known as tauopathies. The most common tauopathy is Alzheimer′s disease, where hyperphosphorylated tau protein forms neurofibrillary tangles inside neurons. Other tauopathies include frontotemporal dementia, progressive supranuclear palsy, and cortico-basal degeneration. These diseases are characterized by cognitive decline, motor disturbances, and memory loss.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service