Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABN226

Sigma-Aldrich

Anti-DENR Antibody, clone 10D9.1

clone 10D9.1, from mouse

Synonym(s):

Density-regulated protein, DRP, Protein DRP1, Smooth muscle cell-associated protein 3, SMAP-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10D9.1, monoclonal

species reactivity

rat, human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DENR(8562)

General description

The protein named density-regulated protein (DRP1), also known as DRP, or Protein DRP1 or Smooth muscle cell-associated protein 3 (SMAP-3), and encoded by the gene DENR/DRP1, is an important translational control protein that appears to function by scanning target mRNAs for the recognition codon when complexed with the translational initiation complex. DRP1 also modulates the translation of particular cancer-related mRNAs when recruited to the translational complex by the oncogene MCTS1 and DRP1 is up regulated in many cancers such as ovarian and breast cancers. Additionally, DRP1’s expression is regulated by increasing cell density through interaction with heterogeneous nuclear ribonucleoprotein D (HNRNPD) protein, a critical protein that complexes with heterogeneous nuclear RNA and pre-mRNAs and fosters mRNA stability. Density-regulated protein (DRP1) is localized to the in heart, skeletal muscle, brain, placenta, liver and pancreas, with weaker expression seen in lung and kidney.

Immunogen

GST-tagged recombinant protein corresponding to the C-terminus of human DENR.

Application

Anti-DENR Antibody, clone 10D9.1 is a mouse monoclonal antibody, validated for use in western blotting & IHC.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected DENR in human cerebral cortex and human cardiac myocytes tissue.

Quality

Evaluated by Western Blotting in PC-12 cell lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected DENR in 10 µg of PC-12 cell lysate.

Target description

~22 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service