MABF3232
Anti-BAFF-R/CD268 Antibody, clone 11C1

Synonym(s):
B-cell-activating factor receptor, BAFF receptor (BAFF-R), BAFFR, BLyS receptor 3, CD268, Tumor necrosis factor receptor superfamily member 13C
About This Item
biological source
mouse
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
11C1, monoclonal
mol wt
calculated mol wt 18.86 kDa
observed mol wt ~N/A kDa
purified by
using protein G
species reactivity
human
packaging
antibody small pack of 100
technique(s)
flow cytometry: suitable
immunohistochemistry: suitable
isotype
IgG1κ
epitope sequence
Unknown
Protein ID accession no.
UniProt accession no.
storage temp.
2-8°C
Gene Information
human ... TNFRSF13C(115650)
Specificity
Immunogen
Application
Evaluated by Immunohistochemistry in human tonsil tissue sections.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected CD268/BAFF-R in human tonsil tissue sections.
Tested Applications
Immunohistochemistry Analysis: A representative lot detected BAFF-R/CD268 in Immunohistochemistry application (Ng, L.G., et al. (2004). J Immunol. 173(2):807-17).
Flow Cytometry Analysis: A representative lot detected BAFF-R/CD268 in Flow Cytometry application (Ng, L.G., et al. (2004). J Immunol. 173(2):807-17).
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Physical form
Reconstitution
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service