Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF3086

Anti-G-CSF-R/CD114 Antibody, clone LMM741

new

Synonym(s):

CD114, G-CSF receptor, G-CSF-R, Granulocyte colony-stimulating factor receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

LMM741, monoclonal

mol wt

calculated mol wt 92.16 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100

technique(s)

flow cytometry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1κ

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

Gene Information

human ... CSF3R(1441)

Specificity

Clone LMM741 is a mouse monoclonal antibody that specifically detects G-CSF-R/CD114. It targets an epitope within the extracellular domain.

Immunogen

CHO-6A11 cells transfected with full length human G-CSF-R/CD114.

Application

Quality Control Testing

Evaluated by Flow Cytometry in human peripheral blood mononuclear cells (PBMC).

Flow Cytometry Analysis(FC): 1.0 ug from a representative lot detected G-CSF-R/CD114 in human peripheral blood mononuclear cells (PBMC).



Tested Applications

Immunoprecipitation Analysis: A representative lot immunoprecipitated G-CSF-R/CD114 in immunoprecipitation application (Nicholson, S.E., et al. (1994). Proc Natl Acad Sci USA. 91(8):2985-8).

Flow Cytometry Analysis: A representative lot detected G-CSF-R/CD114 in flow cytometry application (Layton, J.E., et al. (1999). J Biol Chem. 274(25):17445-51).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Granulocyte colony-stimulating factor receptor (UniProt: Q99062; also known as G-CSF receptor, G-CSF-R, CD114) is encoded by the CSF3R (also known as CSF3R) gene (Gene ID: 1441) in human. CD114/G-CSF-R belongs to the type I subgroup of the superfamily of cytokine receptors. G-CSF-R mediates granulocyte proliferation and differentiation in both normal and malignant hematopoiesis. G-CSF-R functions as the receptor for granulocyte colony-stimulating factor (G-CSF) and is activated via ligand-induced homodimerization. G-CSF-R is a single-pass type I membrane protein produced with a signal peptide sequence (aa 1-24), the removal of which yields the mature single transmembrane (aa 628-650) receptor with a C-terminal cytoplasmic tail (aa 651-836) and a large N-terminal extracellular domain (aa 25-627) composed of a C2-type Ig-like domain (aa 25-117) and five type-III fibronectin domains (aa 125-623). It is highly expressed in bone marrow cells, placenta, lung, lymphoid tissue and peripheral blood granulocytes. The G-CSF/GCSF-R complex mediates survival, proliferation, and maturation of neutrophils and regulates granulopoiesis under both normal and stress conditions. Cell signaling via G-CSF-R is transduced primarily via the Jak/STAT pathway, but the Ras/Raf/MAP kinase and PKB/Akt pathways are also involved. CSF3R gene mutations are well documented in congenital neutropenia as well as the associated predisposition to myelodysplasia and acute myeloid leukemia myeloproliferative disorders. (Ref.: Nicholson, S.E., et al. (1994). Proc Natl Acad Sci USA. 91(8):2985-8; Layton, J.E., et al. (1999). J Biol Chem. 274(25):17445-51).

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service