MABF3078
Anti-HLA‑DQA1 Antibody, clone SPV-L3
Synonym(s):
DC-1 alpha chain, DC-alpha, HLA class II histocompatibility antigen, DQ alpha 1 chain, HLA-DCA, MHC class II DQA1
About This Item
biological source
mouse
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
SPV-L3, monoclonal
mol wt
calculated mol wt 27.81 kDa
observed mol wt ~N/A kDa
purified by
using protein G
species reactivity
human
packaging
antibody small pack of 100
technique(s)
blocking: suitable
flow cytometry: suitable
immunoprecipitation (IP): suitable
isotype
IgG2aκ
epitope sequence
Unknown
Protein ID accession no.
UniProt accession no.
storage temp.
-10 to -25°C
Gene Information
human ... HLA-DQA1(3117)
Specificity
Immunogen
Application
Evaluated by Flow Cytometry in Human peripheral blood mononuclear cells (PBMC).
Flow Cytometry Analysis: 1.0 µg of this antibody detected HLA-DQA1 in one million Human peripheral blood mononuclear cells (PBMC).
Tested Applications
Immunoprecipitation Analysis: A representative lot detected HLA‑DQA1 in Immunoprecipitation applications (Spits, H., et al. (1984). Eur J Immunol. 14(4):299-304).
Blocking of Intefering Clones: A representative lot blocked the activity of CTL clones JR-2-19 and JR-2-26. (Spits, H., et al. (1984). Eur J Immunol. 14(4):299-304).
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Physical form
Reconstitution
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service