Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF2094

Sigma-Aldrich

Anti-alpha TLR10 Antibody, clone 5C2C5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

antibody form

purified antibody

clone

5C2C5
monoclonal

purified by

using protein G

species reactivity

human, human, mouse

concentration

(Please refer to lot specific datasheet.)

technique(s)

flow cytometry: suitable

isotype

IgG1κ

UniProt accession no.

mode of action

| inhibits

General description

Recombinant fragment corresponding to 455 amino acids from the N-terminal, extracellular domain of human Toll-like receptor 10.

Specificity

Clone 5C2C5 is a mouse monoclonal antibody that detects human Toll-like receptor 10.

Application

Epitope: extracellular domain

Quality

Evaluated by Flow Cytometry in human peripheral blood mononuclear cells (PBMC).

Flow Cytometry Analysis: 2 µg/mL of this antibody detected alpha TLR10 in one million human peripheral blood mononuclear cells (PBMC).

Physical properties

94.56 kDa calculated.

Physical form

Purified mouse monoclonal antibody IgG1 in PBS without azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service