MABD172
Anti-THAP11 Antibody, clone 3F3
ascites fluid, clone 3F3, from mouse
Synonym(s):
THAP domain-containing protein 11
About This Item
biological source
mouse
Quality Level
antibody form
ascites fluid
antibody product type
primary antibodies
clone
3F3, monoclonal
species reactivity
human
technique(s)
flow cytometry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable
isotype
IgG1
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... THAP11(57215)
General description
Immunogen
Application
Immunohistochemistry Analysis: A 1:200-1,000 dilution from a representative lot detected THAP11 in paraffin-embedded colon cancer and ovarian tissues.
Flow Cytometry Analysis: A 1:200-400 dilution from a representative lot detected THAP11 in HeLa cells.
Optimal working dilutions must be determined by end user.
Stem Cell Research
Pluripotent & Early Differentiation
Quality
Western Blotting Analysis: A 1:500-2,000 dilution of this antibody detected THAP11 in HeLa and NTERA-2 cell lysates.
Target description
Physical form
Storage and Stability
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Analysis Note
HeLa and NTERA-2 cell lysates
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
nwg
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service