MABC1109
Anti-Mucin-9 Antibody, clone 7E10
clone 7E10, from mouse
Synonym(s):
Oviduct-specific glycoprotein, Estrogen-dependent oviduct protein, Oviductal glycoprotein, Oviductin
About This Item
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
7E10, monoclonal
species reactivity
human
packaging
antibody small pack of 25 μg
technique(s)
ELISA: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable
isotype
IgG2aκ
NCBI accession no.
UniProt accession no.
shipped in
ambient
target post-translational modification
unmodified
Gene Information
human ... OVGP1(5016)
General description
Specificity
Immunogen
Application
Western Blotting Analysis: A representative lot detected Mucin-9 in Western Blotting applications (Maines-Bandiera, S., et. al. (2010). Int J Gynecol Cancer. 20(1):16-22).
ELISA Analysis: A representative lot detected Mucin-9 in ELISA applications (Maines-Bandiera, S., et. al. (2010). Int J Gynecol Cancer. 20(1):16-22).
Immunohistochemistry Analysis: A representative lot detected Mucin-9 in Immunohistochemistry applications (Maines-Bandiera, S., et. al. (2010). Int J Gynecol Cancer. 20(1):16-22).
Apoptosis & Cancer
Quality
Immunohistochemistry Analysis: A 1:250 dilution of this antibody detected Mucin-9 in human oviduct tissue.
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service