Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB6210

Sigma-Aldrich

Anti-Luteinizing Hormone, clone 090-10226 (1mg) KC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

antibody form

purified antibody

Quality Level

clone

090-10226
1A8.9B3
monoclonal

purified by

(Ammonium Sulfate Precipitation)

species reactivity

human, human, mouse

concentration

(Please refer to the Certificate of Analysis for the lot-specific concentration.)

technique(s)

ELISA: suitable

isotype

IgG1

UniProt accession no.

General description

FUNCTION: SwissProt: P01229 # Promotes spermatogenesis and ovulation by stimulating the testes and ovaries to synthesize steroids.
SIZE: 141 amino acids; 15345 Da
SUBUNIT: Heterodimer of a common alpha chain and a unique beta chain which confers biological specificity to thyrotropin, lutropin, follitropin and gonadotropin.
SUBCELLULAR LOCATION: Secreted.
TISSUE SPECIFICITY: Pituitary gland.
DISEASE:SwissProt: P01229 # Defects in LHB are a cause of hypogonadism [MIM:152780]. Hypogonadism is characterized by infertility and pseudohermaphroditism.
SIMILARITY: SwissProt: P01229 ## Belongs to the glycoprotein hormones subunit beta family.
Human pituitary gland

Specificity

Human luteinizing hormone. Less than 0.01% cross reactivity to hCG and greater than 0.1% cross reactivity to FSH.

AFFINITY CONSTANT:

3.2 x 1010

Application

EIA: sensitivity <1 mIU/mL <br /><br />Optimal working dilutions must be determined by the end user.

Physical form

Purified immunoglobulin. Liquid in 10 mM sodium phosphate, 150 mM NaCl, pH 7.4 with 0.1% sodium azide

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Other Notes

Human pituitary gland

Disclaimer

This product is intended for use by IVD manufacturers.
Contact customer service for assistance: oem.reagents@emdmillipore.com
Minimum order quantity required to qualify. Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service