Select a Size
All Photos(1)
Select a Size
Change View
About This Item
UNSPSC Code:
12352200
eCl@ss:
32160405
NACRES:
NA.25
technique(s)
cell based assay: suitable
Quality Level
UniProt accession no.
Gene Information
rat ... SLC4A2(24780)
General description
FUNCTION: SwissProt: P04920 # Plasma membrane anion exchange protein of wide distribution.
SIZE: 1241 amino acids; 137009 Da
SUBCELLULAR LOCATION: Membrane; Multi-pass membrane protein.
SIMILARITY: SwissProt: P04920 ## Belongs to the anion exchanger (TC 2.A.31) family.
SIZE: 1241 amino acids; 137009 Da
SUBCELLULAR LOCATION: Membrane; Multi-pass membrane protein.
SIMILARITY: SwissProt: P04920 ## Belongs to the anion exchanger (TC 2.A.31) family.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service