Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ADCDT500G11

Mobius® ADC Reactor Single-Use Dip Tube

new

Gold, 500 L, 3/8" tubing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

material

silicone cap
silicone gasket
tube (Non-Print PharMed Tubing)

Quality Level

agency

meets EN 2.6.14
meets ISO 10993-4
meets JP 4.01
4.01

meets USP 661
meets USP 788
meets USP 85
meets USP 87
meets USP 88

sterility

sterile; irradiated (at 25-40 kGy)

product line

Mobius®

parameter

4-60 °C temp. range (39-140 °F)

capacity

500 L

application(s)

pharma/biopharma processes

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

General description

Mobius® ADC Reactor Single-Use Dip Tube for reagent addition. No splashing, no foam reagent addition via plunging dip tubes.

Application

Specifically designed for the key conjugation step to manufacture Antibody Drug Conjugates (ADCs).

Packaging

1x3 dip tubes per package.

Legal Information

MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany

related product


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service