Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABN266

Sigma-Aldrich

Anti-KIF5A Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Kinesin heavy chain isoform 5, neuronal kinesin heavy chain, NKHC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KIF5A(3798)

General description

Kinesin heavy chain isoform 5A (KIF5) is a member of the kinesin-like protein family. These proteins are microtubule-dependent motors that slide along microtubules and transport cellular organelles and mRNA to different parts of the cell. In neurons, KIF5A performs this role in an anterograde fashion from the neuronal cell body to axonal or dendritic terminals. KIF5A was initially differentiated from the ubiquitously expressed kinesin as being highly enriched in subsets of neurons and selectively concentrated in the cell body. Mutations in one of the isoforms of KIF5 (KIF5A) is thought to be involved in hereditary spastic paraplegias.

Specificity

This antibody can recognize isoforms KIF5A, KIF5B, and KIF5C.

Immunogen

KLH-conjugated linear peptide corresponding to human KIF5A.

Application

Detect the KIF5A protein using this Anti-KIF5A validated for use in WB & ICC.
Immunocytochemistry Analysis: 5 µg/mL from a representative lot detected KIF5A in K562 cells.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blot in mouse brain nuclear extract.

Western Blot Analysis: 1 µg/mL of this antibody detected KIF5A in 15 µg/mL of mouse brain nuclear extract.

Target description

~117 kDa observed. Uncharacterized bands may appear at ~45 kDa and ~57 kDa in some lysates.

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with 0.02% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse brain nuclear extract

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service