ABC519
Anti-FANCF Antibody
serum, from rabbit
Synonym(s):
Fanconi anemia group F protein, Protein FACF, FANCF
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Recommended Products
biological source
rabbit
Quality Level
antibody form
serum
antibody product type
primary antibodies
clone
polyclonal
species reactivity
human
technique(s)
immunocytochemistry: suitable
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
dry ice
target post-translational modification
unmodified
Gene Information
human ... FANCF(2188)
General description
Fanconi anemia group F protein (FANCF), also known as Protein FACF, and encoded by the gene symbol FANCF, is a DNA repair protein that may operate in a postreplication DNA repair or may be involved in a cell cycle checkpoint function. Recent studies have implicated that FACF may play an important role in interstrand DNA cross-link repair and in the maintenance of normal chromosome stability. Mutations affecting the FANCF gene have been associated with Fanconi anemia complementation group F (FANCF), a disorder affecting all bone marrow elements and resulting in anemia, leukopenia and thrombopenia. This disorder is also associated with cardiac, renal and limb malformations, dermal pigmentary changes, and a predisposition to the development of malignancies. At the cellular level it is associated with hypersensitivity to DNA-damaging agents, chromosomal instability (increased chromosome breakage) and defective DNA repair.
Immunogen
GST-tagged recombinant protein corresponding to human FANCF.
Application
Anti-FANCF is an antibody against FANCF for use in WB, ICC.
Research Category
Apoptosis & Cancer
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
Apoptosis - Additional
Western Blotting Analysis: A representative lot detected FANCF in 293T cells were cotransfected with HA-tagged HES1 and FA coding vectors, HeLa cell extracts treated with L685,458 , 293T cells were cotransfected with FA gene expression plasmids as indicated with either empty, or BACE1 or HES1 coding vectors, 293T cells were cotransfected with FANCF expression plasmids along with increasing amounts of HES1 coding vector (Tremblay, C.S., et al. (2008). Blood Journal. 112:P2062-2070).
Immunocytochemistry Analysis: A representative lot detected FANCF in HeLa cells treated with the gamma-secretase inhibitor L685,458 (Tremblay, C.S., et al. (2008). Blood Journal. 112:P2062-2070).
Immunocytochemistry Analysis: A representative lot detected FANCF in HeLa cells treated with the gamma-secretase inhibitor L685,458 (Tremblay, C.S., et al. (2008). Blood Journal. 112:P2062-2070).
Quality
Evaluated by Western Blotting in MCF-7 cell lysate.
Western Blotting Analysis: A 1:500 dilution of this antibody detected FANCF in 10 µg of MCF-7 cell lysate.
Western Blotting Analysis: A 1:500 dilution of this antibody detected FANCF in 10 µg of MCF-7 cell lysate.
Target description
~45 kDa observed
Physical form
Rabbit polyclonal serum with 0.05% sodium azide.
Unpurified
Storage and Stability
Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Other Notes
Concentration: Please refer to lot specific datasheet.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Cédric S Tremblay et al.
Blood, 112(5), 2062-2070 (2008-06-14)
Fanconi anemia (FA) proteins are thought to play a role in chromosome stability and repair of DNA cross-links; however, these functions may not fully explain the developmental abnormalities and bone marrow failure that are characteristic of FA individuals. Here we
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service