Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABC503

Sigma-Aldrich

Anti-Cathepsin S

from rabbit, purified by affinity chromatography

Synonym(s):

EC: 3.4.22.27

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... CTSS(1520)

General description

Cathepsin S (Uniprot: P25774; EC: 3.4.22.27) is encoded by the CTSS gene (Gene ID: 1520) in human. Cathepsin S is a lysosomal thiol protease that removes invariant chain from MHC class II molecules. It displays higher activity on ZVal-val-Arg-/-Xaa compounds and less activity on Z-Phe-Arg-/-NHMec. Cathepsin S is secreted with a signal (aa 1-16) and a pro-peptide (aa 17-114) sequence that are cleaved to produce active enzyme (aa 115-331). Cathepsin S is involved in antigen presentation, general protein catabolism, and tumor progression. Because of its redistribution or increased levels in human and animal tumors, cathepsin S plays a role in invasion and metastasis. Inhibition of cathepsin S is shown to induce autophagy in tumor cells through EGFR signaling pathway by facilitating EGF-mediated EGFR degradation. Cathepsin S inhibition is shown to delay EGFR degradation and cause EGFR accumulation in the late endosomes at the perinuclear region. (Ref.: Beers, C et al. (2005). J. Immunol. 174(3): 1205-1212; Huang CC et al. (2016). Sci. Rep. 6:29256).

Specificity

This polyclonal antibody detects cathepsin S in human cells. It targets an epitope within 21 amino acids from the internal region.

Immunogen

KLH-conjugated linear peptide corresponding to 21 amino acids from the internal region of human cathepsin S.

Application

Anti-Cathepsin S. Cat. No. ABC503, is a rabbit polyclonal antibody that detects Cathepsin S and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected Cathepsin S in human lymphoma, human small intestine, and human pancreas tissues.
Research Category
Apoptosis & Cancer

Quality

Evaluated by Western Blotting in human testis tissue lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected Cathepsin S in 10 µg of human testis tissue lysate.

Target description

~37 kDa observed; 37.50 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service