Recommended Products
manufacturer/tradename
Novagen®
Quality Level
storage condition
OK to freeze
shipped in
wet ice
storage temp.
2-8°C
General description
The BacMagic-2 DNA Kit provides a positive selection for creating baculoviruses recombinants in preparation for generating recombinant protein expression in insect cells. The kit improves on the traditional method for generating recombinant baculoviruses by eliminating the tedious, time-consuming steps of plaque purification. Like BacMagic DNA, BacMagic-2 DNA is derived from an AcNPV genome, contains an ORF 1629 deletion which prevents nonrecombinant virus from replicating in insect cells, and contains a deletion of the nonessential chitinase (chiA) gene, to improve production of secreted and membrane-targeted proteins. BacMagic-2 DNA have an additional deletion of the v-cath gene, a cathespsin-like cysteine protease, resulting in significantly improved quality and yield for most target proteins. The BacMagic-2 DNA Kit contains the BacMagic-2 DNA, Insect GeneJuice® Transfection Reagent, and a positive control plasmid to monitor co-transfection efficiency.
Not available for sale in Japan
Not available for sale in Japan
Components
- 0.5 µg: BacMagic-2 DNA
- 50 µl: Insect GeneJuice Transfection Reagent
- 2 µg :Transfection Control Plasmid
Warning
Toxicity: Multiple Toxicity Values, refer to MSDS
Legal Information
GENEJUICE is a registered trademark of Merck KGaA, Darmstadt, Germany
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service