Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

550601

Sigma-Aldrich

Pyruvate Kinase Activator

The Pyruvate Kinase Activator controls the biological activity of Pyruvate Kinase. This small molecule/inhibitor is primarily used for Activators/Inducers applications.

Synonym(s):

Pyruvate Kinase Activator, PKM2 Activator I, N-(4-Chloro-3-fluorophenyl)-7-fluoro-2-oxo-1,2,3,4-tetrahydroquinoline-6-sulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11ClF2N2O3S
CAS Number:
Molecular Weight:
372.77
MDL number:
UNSPSC Code:
41106200
NACRES:
NA.77

Quality Level

assay

≥98% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL, clear, colorless

shipped in

ambient

storage temp.

2-8°C

SMILES string

FC1=CC2=C(CCC(N2)=O)C=C1S(NC3=CC=C(F)C(Cl)=C3)(=O)=O

InChI

1S/C15H11ClF2N2O3S/c16-10-6-9(2-3-11(10)17)20-24(22,23)14-5-8-1-4-15(21)19-13(8)7-12(14)18/h2-3,5-7,20H,1,4H2,(H,19,21)

InChI key

VLIHQHQPGAWJCM-UHFFFAOYSA-N

General description

A cell-permeable (Papp x 106 = 9.38 and 7.91 cm/s, respectively, for "A-B" and "B-A" in Caco-2 permeability assay) and biologically stable (t1/2 = 277.2 and 117.5 min., respectively, using human and mouse liver microsome preparation) tetrahydroquinoline-sulfonamide that enhances the donor PEP (phosphoenolpyruvate), but not the acceptor ADP, substrate affinity toward PK (pyruvate kinase) isoform PKM2 (Km = 0.4 vs. 1.9 mM with or without 10 µM activator) and acts as a potent PKM2-selective activator (AC50 = 90 nM using rhPKM2; ACmax = 88%), displaying much reduced activity toward PKL (≤25% at 57 µM) and no effect toward PKM1 or PKR.
A cell-permeable and biologically stable (t1/2 = 277.2 and 117.5 min., respectively, in human and mouse liver microsome preparation) tetrahydroquinoline-sulfonamide that enhances the donor PEP, but not the acceptor ADP, substrate affinity toward pyruvate kinase isoform PKM2 (Km = 0.4 vs. 1.9 mM with or without 10 µM activator) and acts as a potent PKM2-selective activator (AC50 = 90 nM using rhPKM2; ACmax = 88%), displaying much reduced activity toward PKL (≤25% at 57 µM) and no effect toward PKM1 or PKR.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Walsh, M.J., et al. 2011. Bioorg. Med. Chem. Lett.21, 6322.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service