Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

373400

Sigma-Aldrich

Hh/Gli Antagonist, GANT58

The Hh/Gli Antagonist, GANT58, also referenced under CAS 64048-12-0, controls the biological activity of Hh/Gli. This small molecule/inhibitor is primarily used for Cell Signaling applications.

Synonym(s):

Hh/Gli Antagonist, GANT58, 4-(2,4,5-Tri(pyridin-4-yl)thiophen-3-yl)pyridine, NSC 75503

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H16N4S
CAS Number:
Molecular Weight:
392.48
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[s]1c(c(c(c1c5ccncc5)c4ccncc4)c3ccncc3)c2ccncc2

InChI

1S/C24H16N4S/c1-9-25-10-2-17(1)21-22(18-3-11-26-12-4-18)24(20-7-15-28-16-8-20)29-23(21)19-5-13-27-14-6-19/h1-16H

InChI key

USWLOKMMUTWFMD-UHFFFAOYSA-N

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service