Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14-922-M

Sigma-Aldrich

TrkC, active

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

purified by

(Purified using glutathione agarose followed by gel filtration.)

technique(s)

activity assay: suitable

UniProt accession no.

Gene Information

human ... NTRK3(4916)

General description

TrkC is a member of the neurotrophic tyrosine receptor kinase (NTRK) family. This kinase membrane-bound receptor that, upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. Signalling through this kinase leads to cell differentiation and may play a role in the development of proprioceptive neurons that sense
body position. Mutations in this gene have been associated with medulloblastomas, secretory breast carcinomas and other cancers. Several transcript variants encoding different isoforms have been found for this gene.

Packaging

Also available in 10μg size as catalog # 14-922 and in bulk as catalog # 14-922-K.
Also available in 250μg size as catalog # 14-922M and in bulk as catalog # 14-922-K.

Quality

Routinely evaluated by phosphorylation of GEEPLYWSFPAKKK

Physical properties

64 kDa

Physical form

50mM Tris/HCl pH7.5, 300mM NaCl, 0.1mM EGTA, 0.03% Brij-35, 270mM sucrose, 1mM benzamidine, 0.2mM PMSF, 0.1% 2-mercaptoethanol

Storage and Stability

6 months at -70°C

Analysis Note

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service