03-241
RIPAb+ PUM2 Antibody
from rabbit, purified by affinity chromatography
Synonym(s):
pumilio homolog 2 (Drosophila), pumilio homolog 2, pumilio (Drosphila) homolog 2
About This Item
Recommended Products
biological source
rabbit
Quality Level
clone
polyclonal
purified by
affinity chromatography
species reactivity
human, rhesus macaque, mouse
species reactivity (predicted by homology)
chimpanzee (based on 100% sequence homology), rhesus monkey (based on 100% sequence homology), canine (based on 100% sequence homology)
manufacturer/tradename
RIPAb+
Upstate®
technique(s)
RIP: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable
isotype
IgG2a
NCBI accession no.
UniProt accession no.
shipped in
dry ice
Gene Information
human ... PUM2(23369)
General description
PUM2 is a sequence-specific RNA-binding protein that regulates translation and mRNA stability by binding to the 3′-UTR of mRNA targets. The PUM class of proteins plays an active role in the sustainability of mitotic stem cell proliferation and the regulation of cell development and differentiation.
Specificity
Immunogen
Application
Stem Cell Research
RNA Metabolism & Binding Proteins
Representative lot data.
Mouse embryonic stem cell lysate was resolved by electrophoresis, transferred to PVDF membranes and probed with Anti-Pumilio 2 (PUM2) 1 µg/mL. Proteins were visualized using a Donkey Anti-Rabbit IgG conjugated to HRP and chemiluminescence detection system.
Arrow indicates PUM2 (~120 kDa).
Note: A non-specific band may be observed at ~48 kDa in some lysates. (Figure 2).
Immunohistochemistry (Parafin) Analysis:
Representative lot data.
Paraffin-embedded human testis (Fig. 3A) and human teratoma (Fig. 3B) tissues were prepared using heat-induced epitope retrieval in citrate buffer, pH 6.0. Immunostaining was performed using a 1:400 dilution of Anti-Pumilio 2 (PUM2). Reactivity was detected using the IHC-Select Detection Kit (Cat. No. DAB050). Staining pattern appears to be restricted to primary spermatocytes with no Sertoli cell involvement, as expected.
Immunocytochemistry Analysis:
Representative lot data.
Confocal fluorescent analysis of mouse and human embryonic stem cells using Anti-Pumilio 2 (PUM2) (Red). Actin filaments have been labeled with Alexa Fluor™ 488 dye - Phalloidin (Green). Nucleus is stained with DAPI (Blue). (Figure 4) This antibody positively stains the nucleus and cytosol.
Packaging
Quality
Representative lot data.
RIP Lysate prepared from HeLa cells (2 X 10E7 cell equivalents per IP) were subjected to immunoprecipitation using 5 µg of either a normal rabbit IgG or 5 µg of Anti-PUM2 antibody and the Magna RIP® RNA Binding Protein Immunoprecipitation Kit (Cat. # 17-700).
Successful immunoprecipitation of PUM2-associated RNA was verified by qPCR using RIP Primers, NFKBIA mRNA, (Figure 1).
Please refer to the Magna RIP (Cat. # 17-700) or EZ-Magna RIP (Cat. # 17-701) protocol for experimental details.
Target description
Non specific band may be observed at 48 kDa
Physical form
Anti-PUM2 (Rabbit Polyclonal). One vial containing 50 µg of affinity purified polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl, with 0.05% sodium azide, before the addition of 30% glycerol. Store at -20°C.
Concentration: 0.7 mg/mL
Normal Rabbit IgG. One vial containing 125 µg Rabbit IgG in 125 µL storage buffer containing 0.05% sodium azide. Store at -20°C.
RIP Primers, NFKBIA mRNA. One vial containing 75 μL of each primer (5 μM) specific for a region of the human NFKBIA mRNA. Store at -20°C.
FOR: GCT ATT CTC CCT ACC AGC TC
REV: CAG TCA TCA TAG GGC AGC TC
Storage and Stability
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.
Analysis Note
Includes normal rabbit IgG and primers specific for human NFKBIA mRNA.
Other Notes
Legal Information
Disclaimer
Storage Class
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service