Key Documents
CH0441F
Eisco Labs
Eisco Labs Class A Volumetric Flask With Stopper and Graduations
Volumetric Flask, 500ml - Class A, ASTM,Glass Stopper,Blue Graduation
Synonym(s):
Class A, Flasks, Glassware
About This Item
PDF Icon
Shipped & Sold By Eisco Labs
A Trusted Marketplace Seller
PDF Icon
Have a question?
manufacturer/tradename
Eisco Labs
General description
  • 500ml class A borosilicate glass volumetric flask
  • Tolerance ±0.200 ml
  • Comes with solid glass No. 19 stopper
  • 11.5" height and 3" flat base
  • In compliance with ASTM-E288
Borosilicate glass volumetric flask, fitted with a solid glass stopper, in compliance with ASTM-E288 standard. 11.5" Height with a 3" flat base. Capacity 500 ml, Tolerance ±0.200 ml, Stopper No. 19
Eisco Labs Borosilicate glass has the following heat tolerances: Strain Point -515°C | Annealing Point 565°C | Softening Point 820°C.
Calibrated to contain.
Choose from one of the most recent versions:
Certificates of Analysis (COA)