Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

Z317551

Duran® vacuum desiccator plate

porcelain plate, I.D. 150 mm

Synonym(s):

297255707

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43
material:
porcelain plate

material

porcelain plate

manufacturer/tradename

DURAN®
Schott

availability

not available in USA and Canada

I.D.

150 mm

Looking for similar products? Visit Product Comparison Guide

General description

Perforated plate for vacuum desiccators.

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service