GF02149231
T1 - thermocouple alloy, Ni90Cr10
insulated wire, 20m length, 0.05mm conductor diameter, 0.005mm insulation thickness, polyurethane insulation
Synonym(s):
NI095821
Sign Into View Organizational & Contract Pricing
Select a Size
All Photos(1)
Select a Size
Change View
About This Item
UNSPSC Code:
11101711
NACRES:
NA.23
description
polyurethane insulation
thermocouple wire
form
wire (insulated)
manufacturer/tradename
Goodfellow 021-492-31
insulation thickness
0.005 mm
wire L × diam.
20 m × 0.05 mm , conductor diameter
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Application
may be used as one component of a thermocouple
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service