Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

696048

Sigma-Aldrich

Ethyl acetoacetate-1-13C

99 atom % 13C, packaged on demand

Synonym(s):

Acetoacetic ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH213COOC2H5
CAS Number:
Molecular Weight:
131.13
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

Product Name

Ethyl acetoacetate-1-13C, 99 atom % 13C

isotopic purity

99 atom % 13C

assay

99% (CP)

form

liquid

bp

181 °C (lit.)

mp

-43 °C (lit.)

density

1.037 g/mL at 20 °C

mass shift

M+1

SMILES string

CCO[13C](=O)CC(C)=O

InChI

1S/C6H10O3/c1-3-9-6(8)4-5(2)7/h3-4H2,1-2H3/i6+1

InChI key

XYIBRDXRRQCHLP-PTQBSOBMSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

164.3 °F - closed cup

flash_point_c

73.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service