Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

609161

Sigma-Aldrich

Boc-Lys(Z)-OH-α-15N

98 atom % 15N

Synonym(s):

L-Lysine-α-15N, ε−Ν-Cbz derivative

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(Cbz)NH(CH2)4CH(15NH-Boc)CO2H
Molecular Weight:
381.43
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

isotopic purity

98 atom % 15N

technique(s)

bio NMR: suitable

mp

76-79 °C (lit.)

application(s)

peptide synthesis

functional group

Boc

mass shift

M+1

SMILES string

CC(C)(C)OC(=O)[15NH][C@@H](CCCCNC(=O)OCc1ccccc1)C(O)=O

InChI

1S/C19H28N2O6/c1-19(2,3)27-18(25)21-15(16(22)23)11-7-8-12-20-17(24)26-13-14-9-5-4-6-10-14/h4-6,9-10,15H,7-8,11-13H2,1-3H3,(H,20,24)(H,21,25)(H,22,23)/t15-/m0/s1/i21+1

InChI key

BDHUTRNYBGWPBL-OVLDBCRESA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service