Key Documents
AATB-11119
AAT Bioquest
AAT Bioquest Amplite® Colorimetric Bradford Protein Quantitation Assay Kit
5000 Tests
About This Item
PDF Icon
Shipped & Sold By AAT Bioquest
A Trusted Marketplace Seller
PDF Icon
Have a question?
manufacturer/tradename
AAT Bioquest
General description
The traditional Bradford protein assay is widely used for quantifying protein concentrations. Amplite®™ Colorimetric Braford Protein Quantitation Assay Kit is a two-component assay to determine total protein concentrations. The assay is based on the same Coomassie Blue G-250 protein indicator as Bradford protein assay and provides comparable accuracy. Our proprietary formulation makes our kit much more convenient and rapid. The protein signal is monitored around 600 nm and assay is completed within 30 minutes.
Components:
  • Bradford Assay Solution-18 mL
  • BSA Standard (1 mg/mL)-1 mL
Amplite® Colorimetric Bradford Protein Quantitation Assay Kit is manufactured by AAT Bioquest
Legal Information
Amplite® is a trademark of AAT Bioquest
Choose from one of the most recent versions:
Certificates of Analysis (COA)