Skip to Content
Merck

809365

Sigma-Aldrich

Lithium hexafluorophosphate solution

in ethylene carbonate and ethylmethyl carbonate, 2.0 M LiPF6 EC/EMC=50/50(v/v), battery grade

Synonym(s):

2 M LiPF6 [EC/EMC=50/50(v/v)]

Sign Into View Organizational & Contract Pricing

About This Item

Linear Formula:
LiPF6
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.23
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

battery grade

form

liquid

density

1.2966 g/mL

application(s)

battery manufacturing

SMILES string

F[P-](F)(F)(F)(F)F.[Li+]

InChI

1S/F6P.Li/c1-7(2,3,4,5)6;/q-1;+1

InChI key

AXPLOJNSKRXQPA-UHFFFAOYSA-N

General description

Battery grade: H2O <15 ppm, HF <50 ppm, and APHA <50 ppm.
Lithium hexafluorophosphate solution in ethylene carbonate and ethylmethyl carbonate is a class of electrolytic solution that can be used in the fabrication of lithium-ion batteries. Lithium-ion batteries consist of anode, cathode, and electrolyte with a charge-discharge cycle. These materials enable the formation of greener and sustainable batteries for electrical energy storage.

Application

The solutions of LiPF6 in alkyl carbonates find applications as electrolytes in lithium ion batteries and the use of high quality battery grade electrolytes having extremely low water (<15 ppm) and hydrogen fluoride (<50 ppm) contents are critical for achieving high electrochemical performance.

Preparation Note

  • Do not use any glass equipment.
  • Please handle under inert and moisture free environment.

Legal Information

Product of MU Ionic Solutions Corp


Still not finding the right product?


Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT RE 2 Oral

Target Organs

Bone,Teeth, Kidney

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

80.1 °F

Flash Point(C)

26.7 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library